Learn More
Hamilton™ DIN Liq-Glass Temp Electrode
Product Code. 11560302
Click to view available options
Length (Metric):
120 mm
Unit Size:
Each
Product Code. 11560302
Brand: Hamilton™ 238406
Only null left
Specifications
Accuracy (mV) 0 ± 20mV
Accuracy (Temperature) 0°C to 60°C
Diameter (Metric) 12 mm
Electrical Requirements 0 ± 20 mV, 57 to 59 mV
Length (Metric) 120 mm
pH Range 0 to 14
Temperature (Metric) Range 0°C to 60°C
Certifications/Compliance Yes, with measured values in Buffer pH 4.01 and pH 7.00