missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dinitrophenyl-KLH Polyclonal Antibody
GREENER_CHOICE

Product Code. 10104292
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10104292 500 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10104292 Supplier Invitrogen™ Supplier No. A6430

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Dinitrophenyl-KLH Polyclonal Antibody for Flow, ICC/IF, IHC

DNP-KLH antibodies are used for detecting DNP-labeled molecules. Fluorescent and biotinylated anti-DNP-KLH conjugates can serve as second-step reagents for signal amplification in protein and nucleic acid detection systems, including in situ hybridization studies employing DNP-modified DNA probes. Because anti-DNP antibody binds well to DAMP (D-1552), an acidotropic reagent that can penetrate cell membranes, anti-DNP-KLH antibodies can also be used with DAMP for immunochemical localization of intracellular compartments with low pH. DNP-KLH antibodies may recognize compounds with a trinitrophenyl group, allowing the isolation of cell-surface molecules of human melanoma cells that have been labeled with trinitrobenzene sulfonic acid (TNBS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dinitrophenyl-KLH
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5mM sodium azide; pH 7.2
Gene Alias C6H3N2O4; DNP-KLH
Host Species Rabbit
Immunogen Dinitrophenyl-KLH.
Purification Method Purified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.