Learn More
Dinotefuran, MedChemExpress
Product Code. 16498466
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498466 100 mg 100mg
16474497 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16498466 Supplier Medchem Express Supplier No. HYB0827
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress Dinotefuran is an insecticide of the neonicotinoid class, its mechanism of action involves disruption of the insect's nervous system by inhibiting nicotinic acetylcholine receptors.
Dinotefuran is an insecticide of the neonicotinoid class, its mechanism of action involves disruption of the insect's nervous system by inhibiting nicotinic acetylcholine receptors.
Tekniske data
Chemical Name or Material Dinotefuran
CAS 165252-70-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C7H14N4O3
Quantity 100 mg
Synonym MTI-446
Solubility Information H2O : 50 mg/mL (247.27 mM; Need ultrasonic)
SMILES O=[N+](N/C(NC)=N/CC1COCC1)[O-]
Molecular Weight (g/mol) 202.21
Formula Weight 202.21
Percent Purity 98.88%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Vis mere Vis mindre
Research purposes only
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.