Learn More
Thermo Scientific™ Dionex™ Fused Silica Tubing
Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric) Inner:
10 μm
20 μm
25 μm
30 μm
50 μm
75 μm
100 μm
150 μm
200 μm
Length (Metric):
2.5 m
5 m
Unit Size:
Each
Description
Thermo Scientific™ Fused Silica Tubing provides excellent chemical durability and inertness.
Specifications
Specifications
| For Use With (Equipment) | LC-Packings Ultimate Capillary/Nano HPLC |
| Material | Fused Silica |
| Type | Capillary Tubing |
| Length (Metric) | 5 m |
| Diameter (Metric) Inner | 150 μm |
| Diameter (Metric) Outer | 280 μm |
| Product Line | nanoLC |