missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIP13B (APPL2) Monoclonal Antibody (OTI8F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF812391
This item is not returnable.
View return policy
Description
DIP13B (APPL2) Monoclonal antibody specifically detects DIP13B (APPL2) in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| DIP13B (APPL2) | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8K3G9, Q8NEU8 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 216190, 362860, 55198 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI8F1 | |
| Unconjugated | |
| APPL2 | |
| adapter protein containing PH domain, PTB domain and leucine zipper motif 2; adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2; adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 2; Appl2; DCC-interacting protein 13-beta; DIP13 beta; DIP13B; dip13-beta; Dip3 beta; Dip3b; RGD1563028 | |
| Full length human recombinant protein of human APPL2 (NP_060641) produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction