missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DIP2B Polyclonal Antibody
GREENER_CHOICE

Product Code. 13279568
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13279568 100 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13279568 Supplier Invitrogen™ Supplier No. PA522089

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, Molt-4, mouse brain. Predicted reactivity: Mouse (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the disco-interacting protein homolog 2 protein family. The encoded protein contains a binding site for the transcriptional regulator DNA methyltransferase 1 associated protein 1 as well as AMP-binding sites. The presence of these sites suggests that the encoded protein may participate in DNA methylation. This gene is located near a folate-sensitive fragile site, and CGG-repeat expansion in the promoter of this gene which affects transcription has been detected in individuals containing this fragile site on chromosome 12.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIP2B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene DIP2B
Gene Accession No. Q3UH60, Q9P265
Gene Alias 4932422C22; AI317237; AI854602; DIP2 disco-interacting protein 2 homolog B; DIP2 disco-interacting protein 2 homolog B (Drosophila); DIP2 homolog B; Dip2b; disco interacting protein 2 homolog B; disco-interacting protein 2 homolog B; KIAA1463; mKIAA1463
Gene Symbols DIP2B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human DIP2B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239667, 57609
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.