missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIP2C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35826-100ul
This item is not returnable.
View return policy
Description
DIP2C Polyclonal antibody specifically detects DIP2C in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| DIP2C | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| DIP2 disco-interacting protein 2 homolog C (Drosophila), disco-interacting protein 2 homolog C, FLJ34444, FLJ44075, KIAA0934DIP2 homolog C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-260 of human DIP2C (NP_055789.1).,, Sequence:, LAKHKERKMAVPMPSKRRSLVVQTSMDAYTPPDTSSGSEDEGSVQGDSQGTPTSSQGSINMEHWISQAIHGSTTSTTSSSSTQSGGSGAAHRLADVMAQTHIENHSAPPDVTTYTSEHSIQVERPQGSTGSRTAPKYGNAELMETGDGVPVSSRVSAKIQQ | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 22982 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction