missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DIPA Polyclonal Antibody
GREENER_CHOICE

Product Code. 13290339
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13290339 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13290339 Supplier Invitrogen™ Supplier No. PA526129

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology.

DIPA functions as a transcriptional repressor. It may inhibit the activity of CTNNB1 in a TP53-dependent manner and thus regulate cell growth. It may function in adipocyte differentiation, negatively regulating mitotic clonal expansion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIPA
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CCDC85B
Gene Accession No. Q15834
Gene Alias AI842788; BB115872; CCDC85B; coiled-coil domain containing 85B; coiled-coil domain-containing protein 85B; Delta-interacting protein A; delta-interacting protein A homolog; Dipa; hepatitis delta antigen interacting protein A; hepatitis delta antigen-interacting protein A; Hepatitis delta antigen-interacting protein A homolog
Gene Symbols CCDC85B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 69-96 amino acids from the central region of human DIPA.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11007
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.