missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenhydramine hydrochloride, MedChemExpress
Description
Diphenhydramine hydrochloride is a first-generation histamine H1-receptor antagonist with anti-cholinergic effect. Diphenhydramine hydrochloride can across the ovine blood-brain barrier (BBB).
Specifications
Specifications
| Chemical Name or Material | Diphenhydramine hydrochloride |
| Physical Form | Solid |
| CAS | 147-24-0 |
| Color | White |
| Quantity | 250 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C17H22ClNO |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : 100 mg/mL (342.68 mM; Need ultrasonic) ∣H2O : 100 mg/mL (342.68 mM; Need ultrasonic) |
| SMILES | CN(C)CCOC(C1=CC=CC=C1)C2=CC=CC=C2.Cl |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?