missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl disulfide, 98%

Product Code. p-7020580
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11479010

Brand: Thermo Scientific Alfa Aesar A12586.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 882-33-7 | C12H10S2 | 218.332 g/mol

Diphenyl disulfide is one of the most popular organic disulfides used in organic synthesis. It is used as a reagent for the α-phenylsulfenylation of carbonyl compounds. It participates in hydrothiolation of alkynes via amine-mediated single electron transfer mechanism. It is the hydrolysis product of dyfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl disulfide is one of the most popular organic disulfides used in organic synthesis. It is used as a reagent for the α-phenylsulfenylation of carbonyl compounds. It participates in hydrothiolation of alkynes via amine-mediated single electron transfer mechanism. It is the hydrolysis product of dyfonate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882-33-7
Molecular Formula C12H10S2
Molecular Weight (g/mol) 218.332
MDL Number MFCD00003065
InChI Key GUUVPOWQJOLRAS-UHFFFAOYSA-N
Synonym diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene
PubChem CID 13436
IUPAC Name (phenyldisulfanyl)benzene
SMILES C1=CC=C(C=C1)SSC2=CC=CC=C2

Specifications

Melting Point 58°C to 62°C
Boiling Point 310°C
Odor Unpleasant
Quantity 50 g
Beilstein 639794
Solubility Information Insoluble in water.
Formula Weight 218.34
Percent Purity 98%
Chemical Name or Material Diphenyl disulfide
compliance-icons compliance-icons
Product Identifier
  • Diphenyl disulfide
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Diphenyl disulfide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.