missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Disulfide, TRC

Product Code. 17921895
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17921895 10 g 10g
30134322 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17921895 Supplier Toronto Research Chemicals Supplier No. TRCD48763010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882-33-7
Formula Weight 218.0224
InChI Formula InChI=1S/C12H10S2/c1-3-7-11(8-4-1)13-14-12-9-5-2-6-10-12/h1-10H
IUPAC Name (phenyldisulfanyl)benzene
Molecular Formula C12 H10 S2
Molecular Weight (g/mol) 218.34
Percent Purity >95%
SMILES S(Sc1ccccc1)c2ccccc2
Synonym Phenyl disulfide (8CI), Diphenyl disulfide, (Phenyldisulfanyl)benzene, 1,1'-Dithiodibenzene, 1,2-Diphenyldisulfane, Biphenyl disulfide, DPDS, Diphenyl disulphide, NSC 2689, Diphenyl Disulfide
Chemical Name or Material Diphenyl disulfide
Grade HPLC
Show More Show Less

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.