missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphtheria Toxin Antibody (3B6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-2564
This item is not returnable.
View return policy
Description
Diphtheria Toxin Monoclonal specifically detects Diphtheria Toxin in Bacteria samples. It is validated for Sandwich ELISA.
Specifications
| Diphtheria Toxin | |
| Monoclonal | |
| Unconjugated | |
| Diphtheria Toxin, tox | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Diphtheria toxoid. | |
| 0.2 mg | |
| Virology Bacteria and Parasites | |
| Reacts with Diphtheria toxin and anatoxin. Does not react with free A or B subunits of Diphtheria toxin. | |
| Store at 4C. Do not freeze. | |
| IgG1 |
| ELISA | |
| 3B6 | |
| Sandwich ELISA 1:100-1:2000 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction