missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIRAS2 Antibody (OTI10H1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71967APC
This item is not returnable.
View return policy
Description
DIRAS2 Monoclonal antibody specifically detects DIRAS2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| DIRAS2 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI10H1 | |
| Western Blot | |
| DIRAS family, GTP-binding RAS-like 2, Di-Ras2, Distinct subgroup of the Ras family member 2, DKFZp761C07121, GTP-binding protein Di-Ras2 | |
| Human recombinant protein fragment corresponding to amino acids 1-196 of human DIRAS2(NP_060064) produced in E.coli. | |
| 0.1 mL | |
| Signal Transduction | |
| 54769 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction