Learn More
DIRAS2 Monoclonal Antibody (OTI10G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809448S
This item is not returnable.
View return policy
Description
DIRAS2 Monoclonal antibody specifically detects DIRAS2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| DIRAS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q496Y0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79836 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI10G5 | |
| Unconjugated | |
| LONRF3 | |
| 2900052J15Rik; AI414999; DIRAS family GTP binding RAS like 2; DIRAS family GTPase 2; DIRAS family, GTP-binding RAS-like 2; DIRAS2; Di-Ras2; Distinct subgroup of the Ras family member 2; GTP-binding protein Di-Ras2 | |
| Human recombinant protein fragment corresponding to amino acids 206-456 of human LONRF3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |