Learn More
DIRAS2 Monoclonal Antibody (OTI10G5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809448S
Encompass_Preferred
 View more versions of this product
Product Code. 17541684
  • € 242.00 / 30µL
Description
DIRAS2 Monoclonal antibody specifically detects DIRAS2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
DIRAS2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q496Y0
Mouse
Affinity chromatography
RUO
79836
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Immunohistochemistry (Paraffin), Western Blot
OTI10G5
Unconjugated
LONRF3
2900052J15Rik; AI414999; DIRAS family GTP binding RAS like 2; DIRAS family GTPase 2; DIRAS family, GTP-binding RAS-like 2; DIRAS2; Di-Ras2; Distinct subgroup of the Ras family member 2; GTP-binding protein Di-Ras2
Human recombinant protein fragment corresponding to amino acids 206-456 of human LONRF3 produced in E.coli.
30 μL