missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DIRC1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15896223
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15896223 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15896223 Supplier Invitrogen™ Supplier No. PA567811

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DIRC1 Polyclonal Antibody for Western Blot

This gene consists of two exons and encodes a 104aa protein. Disruption of this gene by translocation t(2;3)(q33;q21) is associated with familial clear cell renal cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIRC1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DIRC1
Gene Accession No. Q969H9
Gene Alias DIRC1; disrupted in renal cancer protein; disrupted in renal carcinoma 1; disrupted in renal carcinoma protein 1
Gene Symbols DIRC1
Host Species Rabbit
Immunogen Synthesized peptide derived from human DIRC1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116093
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.