Learn More
Brady™ Direct Thermal Printable Labels
Product Code. 31751202 Shop All Brady Products
Change view
Click to view available options
Height (English):
6.00 in.
Unit Size:
1 roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (English) Unit Size
31751202 6.00 in. 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31751202 Supplier Brady™ Supplier No. 324907
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Printable, paper labels for direct thermal printers
  • Ideal label sizing for logistical applications.
Specifications
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Matte
Format Die-cut
Material Paper
Printable Sides Single-side Printable
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
For Use With (Equipment) Brady i6100
No. per Pack 3000
Shape Rectangle
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (L x W) 152.40 x 101.60 mm
Length (English) 152.40 mm
Width (English) 4 in.
Width (Metric) 101.60 mm
Width (English) Printable Area 101.60 mm
Height (English) 6.00 in.
Height (Metric) 152.40 mm
Alternate Part Number I61-DT-25-7604-1