Learn More
DIS3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29664-1-AP-20UL
This item is not returnable.
View return policy
Description
DIS3 Polyclonal antibody apecifically detetects DIS3 antigen in Human, Mouse Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
| DIS3 | |
| Polyclonal | |
| Unconjugated | |
| NM_014953 | |
| Rabbit | |
| Antigen affinity purification | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 300 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| DIS3 | |
| DIS3 fusion protein Ag31711 | |
| 20 μL | |
| 22894 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG |