missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DIS3L Polyclonal antibody specifically detects DIS3L in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | DIS3L |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Formulation | PBS (pH 7.0) |
| Gene Alias | DIS3 mitotic control homolog (S. cerevisiae)-like, DIS3-like exonuclease 1, EC 3.1.13, FLJ38088, KIAA1955EC 3.1.13.-, MGC4562 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human DIS3L fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?