missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIS3L2 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81805UV
This item is not returnable.
View return policy
Description
DIS3L2 Polyclonal antibody specifically detects DIS3L2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| DIS3L2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 129563 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| DyLight 350 | |
| DIS3 mitotic control homolog (S. cerevisiae)-like 2, DIS3-like exonuclease 2, FAM6A, member A | |
| DIS3L2 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human DIS3L2. The immunogen is located within amino acids 720 - 770 of DIS3L2. Amino Acid Squence: DTLQKQADHCNDRRMAS | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction