missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DIS3L2 Monoclonal Antibody (6C7B2)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA531718

Product Code. 16305154

  • € 518.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-31718 has been tested in indirect ELISA.

DIS3L2 encodes a protein that is similar in sequence to 3'/5' exonucleolytic subunits of the RNA exosome. The exosome is a large multimeric ribonucleotide complex responsible for degrading various RNA substrates. Several transcript variants, some protein-coding and some not, have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DIS3L2
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
Q8IYB7
DIS3L2
Purified recombinant fragment of human DIS3L2 (AA: 27-250) expressed in E. coli.
100 μL
Primary
Human
Antibody
IgG1
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
6C7B2
Unconjugated
DIS3L2
4930429A22Rik; 8030493P09Rik; DIS3 like 3'-5' exoribonuclease 2; DIS3 mitotic control homolog (S. cerevisiae)-like 2; DIS3 mitotic control homolog-like 2; DIS3 mitotic control-like 2; Dis3l2; DIS3-like 3'-5' exoribonuclease 2; DIS3-like exonuclease 2; FAM6A; family with sequence similarity 6, member A; hDIS3L2; PRLMNS; RGD1560168
Mouse
Protein G
RUO
129563
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.