Learn More
Lenz Laborglasintrumente Disc Knots
Description
- Central bore for bar dia. 22 mm, height adjustable
- With 8 tap holes M12
- Complete with set-screw M12
- Material: steel, chrome plated
- Pack: 1 piece
Specifications
Specifications
| Includes | 8 tap holes |
| Material | Steel, chrome-plated |