Learn More
Lenz Laborglasintrumente Disc Knots
Product Code. 11947091
Change view
Click to view available options
Material:
Steel, chrome-plated
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
11947091 Steel, chrome-plated Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11947091 Supplier Lenz Laborglasintrumente Supplier No. 06247618
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Central bore for bar dia. 22 mm, height adjustable
  • With 8 tap holes M12
  • Complete with set-screw M12
  • Material: steel, chrome plated
  • Pack: 1 piece
Specifications
Includes 8 tap holes
Material Steel, chrome-plated