Learn More
Hirschmann™ Discharge Tube Unit with Luer-Lock Connection
Product Code. 10762951
Change view
Click to view available options
Type:
Discharge Tube Unit
Ejection Tube Unit
Material:
FEP, PFA
FEP, Polypropylene
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Material unitSize
10762951 Ejection Tube Unit FEP, Polypropylene Each
10191521 Discharge Tube Unit FEP, Polypropylene Each
10164351 Ejection Tube Unit FEP, PFA Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10762951 Supplier Hirschmann™ Supplier No. 9313008
Only null left
Add to basket
Add to basket
Dispensing nozzle with FEP-tubing and Luer-Lock-Connection
  • Guided tubing
  • Screw-coupled
  • With Luer-Lock connector for filtration
Specifications
Quantity 1 Each
Type Ejection Tube Unit
For Use With (Equipment) ceramus™ classic and EM-dispenser for 30 + 60 mL
Material FEP, Polypropylene
Description With Luer-Lock-connection