missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dishevelled-2 Antibody (OTI2D11), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70611H
This item is not returnable.
View return policy
Description
Dishevelled-2 Monoclonal specifically detects Dishevelled-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| Dishevelled-2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| DVL2 | |
| Recombinant protein fragment corresponding to amino acids 1-320 of human Dishevelled-2 (NP_004413) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Immunohistochemistry | |
| OTI2D11 | |
| Immunohistochemistry | |
| dishevelled 2 (homologous to Drosophila dsh), dishevelled, dsh homolog 2 (Drosophila), Dishevelled-2, DSH homolog 2, segment polarity protein dishevelled homolog DVL-2 | |
| Mouse | |
| Affinity Purified | |
| Phospho Specific, Signal Transduction, Wnt Signaling Pathway | |
| 1856.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction