missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dishevelled-2 Antibody (OTI2D11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46480
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Immunohistochemistry
Dishevelled-2 Monoclonal specifically detects Dishevelled-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Dishevelled-2 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
| dishevelled 2 (homologous to Drosophila dsh), dishevelled, dsh homolog 2 (Drosophila), Dishevelled-2, DSH homolog 2, segment polarity protein dishevelled homolog DVL-2 | |
| Mouse | |
| 78.8 kDa | |
| 0.1 mL | |
| Signal Transduction, Wnt Signaling Pathway | |
| 1856 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI2D11 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| DVL2 | |
| Human recombinant protein fragment corresponding to amino acids 1-320 of human DVL2(NP_004413) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction