missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disialoganglioside GD2 Mouse anti-Human, BUV661, Clone: 14.G2a, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 749814
This item is not returnable.
View return policy
Description
Disialoganglioside GD2 Monoclonal antibody specifically detects Disialoganglioside GD2 in Human samples. It is validated for Flow Cytometry
Specifications
| Disialoganglioside GD2 | |
| Monoclonal | |
| Brilliant Ultraviolet 661 | |
| GD2; Disialoganglioside GD2; Ganglioside GD2 | |
| LAN-1 human neuroblastoma cells | |
| RUO | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 14.G2a | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction