Learn More
Bode Chemie™ Dismofix™ G Gloss Finish Cleaner
Product Code. 10420543
Change view
Click to view available options
Color:
Yellow
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10420543 Yellow 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10420543 Supplier Bode Chemie™ Supplier No. 9802003
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For the maintenance cleaning of all washable surfaces and coated floors.
  • Dries without leaving streaks
  • Low foaming
  • For streak-free cleaning of sensitive surfaces such as glass, plastics and ceramics
  • For automated cleaning of all treated floors with cleaning machines
  • For the cleaning of conductive flooring
Specifications
Product Type Dismofix G Gloss Finish Cleaner
Form Liquid
pH 7 (at 20°C)
Appearance Yellow
Color Yellow
For Use With (Application) Cleaning
Volume (Metric) 5 L