missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DISP2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81917AF350
This item is not returnable.
View return policy
Description
DISP2 Polyclonal antibody specifically detects DISP2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| DISP2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 350 | |
| Dispatched B, Dispatched Homolog 2 (Drosophila), DISPB, HsT16908, KIAA1742, Protein Dispatched Homolog 2 | |
| Disp2 antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human Disp2. The immunogen is located within amino acids 1210 - 1260 of Disp2. Amino Acid Squence: CPALQTSSPYKQAGPS | |
| 0.1 mL | |
| Signal Transduction | |
| 85455 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction