missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DISP2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81918B
This item is not returnable.
View return policy
Description
DISP2 Polyclonal antibody specifically detects DISP2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| DISP2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| Dispatched B, Dispatched Homolog 2 (Drosophila), DISPB, HsT16908, KIAA1742, Protein Dispatched Homolog 2 | |
| DISP2 antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human DISP2. The immunogen is located within amino acids 110 - 160 of DISP2. Amino Acid Squence: SPSPAPSQRDGTWKPPA | |
| 0.1 mL | |
| Signal Transduction | |
| 85455 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction