Learn More
Witeg™ Dispenser
Click to view available options
Volume (Metric):
1 to 10 mL
500 and 1000 μL
Unit Size:
Each
Specifications
Specifications
| Accuracy | ± 0.06mL |
| Product Type | Bottletop Dispenser |
| Autoclavable | Autoclavable |
| Material | Platinum Spring |
| Volume (Metric) | 1 to 10 mL |