Learn More
BRAND™ Tilt Measure Dispenser with Bottle
Shop All Brand Products
Click to view available options
Volume (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
Unit Size:
Each
Description
- Dispenser head made from Borosilicate glass (Boro 3.3)
- Calibrated to deliver (TD, Ex)
- Inscriptions in green enamel
- Particularly well-suited for dispensing suspensions
- Complete with 1000mL soda-lime glass reservoir bottle with 29/32 neck size
Specifications
Specifications
| Product Type | Top Tilt Pipette Dispenser |
| Color | Clear |
| Material | Borosilicate Glass |
| Volume (Metric) | 100 mL |
| Includes | 1000mL soda-lime glass reserve bottle |