missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Disposable Dropping Pipet

Pipettes made of plastic are particularly suited for training purposes, in industrial labs, during field research and applications where pipettes made of glass cannot be used, e.g. in flourine and silicate chemistry

Brand:  Buerkle™ 7555-0003

27.20 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.



Additional Details : Weight : 0.28000kg

 View more versions of this product

Product Code. 11971531

  • € 29.65 / 1 pack
Estimated Shipment: 10-05-2024
to see stock.
Add to basket

Description

Description

  • LDPE
  • dropping pipette, not graduated
Specifications

Specifications

LDPE
1 Ea.
155 mm
Single-use
Suited for Training Purposes, In Industrial Labs, During Field Research and Applications
2.4 mL
Dropping Pipette
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers