Learn More
Buerkle™ Disposable Dropping Pipet
Pipettes made of plastic are particularly suited for training purposes, in industrial labs, during field research and applications where pipettes made of glass cannot be used, e.g. in flourine and silicate chemistry
€ 29.65
Specifications
Material LDPE
For Use With (Application) Suited for Training Purposes, In Industrial Labs, During Field Research and Applications
Quantity 1 Ea.
Type Dropping Pipette
Disposable Single-use
View More Specs
Products 3
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11961531
View Documents Promotion Details
Buerkle™
7555-0002
1.7 mL
€ 29.65
1 pack
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Add to basket
11951531
View Documents Promotion Details
Buerkle™
7555-0001
1.8 mL
€ 29.65
1 pack
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Add to basket
11971531
View Documents Promotion Details
Buerkle™
7555-0003
2.4 mL
€ 29.65
1 pack
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Add to basket
Description
Description
  • LDPE
  • dropping pipette, not graduated
Specifications
Specifications
LDPE
1 Ea.
Single-use
Suited for Training Purposes, In Industrial Labs, During Field Research and Applications
Dropping Pipette