missing translation for 'onlineSavingsMsg'
Learn More

Terracycle™ Disposable Face Masks Zero Waste Box™

Product Code. 17895735
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17895735 Large Each
17835925 Small Each
17885695 Medium Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17895735 Supplier Terracycle™ Supplier No. CS131LNL
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recycle Disposable Face Masks using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

Use this Zero Waste Box to recycle:

  • Non-woven disposable face masks, plastic face shields and visors, 3-ply surgical masks and dust masks
  • We accept all types of disposable face masks made from plastic, such as dust masks, FFP2 (or so-called ''N95''), FFP3 and surgical masks (as long as they don't come from a medical environment)

Approximate waste units accepted:

  • Small: 430 face masks
  • Medium: 1100 face masks
  • Large: 2200 face masks

Do not send in:

  • Medical waste.
    • This includes material contaminated with blood or bodily fluids and waste that originate from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
  • Fabric, homemade or paper masks or other types of protective garments
  • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) or material contaminated with blood which presents a danger to the environment, or to people
  • Batteries, pressurized canisters, broken glass

Specifications

Capacity (Metric) 25 kg
Capacity (English) 55 lb.
Color Beige, Green, Red, White
Dimensions (L x W x H) 38 x 38 x 97 cm
Disposable Single-use
Latex Free Yes
Size Large
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.