Learn More
Disposable Luer-Slip Two-Part Syringe
Click to view available options
Volume (Metric):
10 to 12 mL
2 to 3 mL
5 to 6 mL
Unit Size:
Pack of 100
Description
- Available from sizes 1 to 50mL (60mL)
- Smooth flow, tight, high transparency barrel
- No rubber, styrene or DEHP
- Latex- and silicone-oil-free
- Pyrogen-free, PVC-free, non-toxic
- Sterile, individually blister strip packed
- Your first choice for an inert syringe
- Defined position of the plunger at volume “0” to feel when plunger is completely inserted
- LDS: no dead space for 1mL syringe
- Extended graduation for wider range of application
- Positive safety stop to prevent accidental spills
- 1 to 20mL (24mL) available also with by-packed needle
- 2 to 20mL available as Oral syringe with purple plunger
- 2mL to 50 mL (60mL) also available with Luer Lock
- 30mL and 50mL also available with Catheter tip
- All sizes available in non-sterile version (bulk) upon request
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | DIN, EN, ISO and FDA standards, Quality management system certified according to ISO 13485:2003 / AC 2007, CE-marked |
| Material | Polypropylene |
| Color | Transparent Barrel |
| Description | Smudge-proof scale |
| Latex Free | Yes |
| Product Type | Luer-slip 2-part Syringe |
| Disposable | Single-use |
| Graduations | 0.2 mL |
| Quantity | 100/Pk. |
| Show More |