Learn More
B Braun™ Injekt™ Solo Single-use Syringe
Product Code. 10641200
Change view
Click to view available options
Capacity (Metric):
3 mL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10641200 3 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10641200 Supplier B Braun™ Supplier No. 9202528
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Made of polypropylene and polyethylene B Braun™ Injekt™ Solo Single-use Syringe includes a green plunger with a highly transparent barrel. „Latex-free/PVC-free „„Silicone oil-free tube easily aspirates up to maximum volume.
2-piece single-use syringe
  • Made of polypropylene/polyethylene
  • „„Green plunger
  • „„Highly transparent barrel „
  • „High contrast black graduation with permanent marking provides ideal readability „„
  • Safe plunger backstop
  • „„Easy aspiration up to maximum volume „„
  • Latex-free/PVC-free „„Silicone oil-free
Specifications
Material Polypropylene, Polyethylene
Sterility Non-sterile
Capacity (Metric) 3 mL
Certifications/Compliance EN ISO 7886-1
Closure Type Centric Luer Cone
Color Green Plunger
Disposable Single-use
Latex Free Yes