Learn More
B Braun™ Dissecting Standard Forceps
Click to view available options
Length (Metric):
105 mm
Unit Size:
Each
Specifications
Specifications
| No. per Pack | 1 Pack |
| Design | Straight geometry |
| Length (Metric) | 105 mm |
| Product Type | Forceps, Dissecting |
| For Use With (Application) | Dental surgery |
| Teeth Count | 1 x 2 |
| Type | Dissecting Standard Forceps |