Learn More
B Braun™ Dissecting Standard Forceps
Product Code. 10074363
Click to view available options
Length (Metric):
105 mm
Unit Size:
Each
Product Code. 10074363
Brand: B Braun™ BD552R
Only null left
Specifications
No. per Pack 1 Pack
Design Straight geometry
Length (Metric) 105 mm
Product Type Forceps, Dissecting
For Use With (Application) Dental surgery
Teeth Count 1 x 2
Type Dissecting Standard Forceps