missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Distemper Virus Antibody (5-4), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-2604F
This item is not returnable.
View return policy
Description
Distemper Virus Monoclonal specifically detects Distemper Virus in Virus samples. It is validated for ELISA, Sandwich ELISA.
Specifications
| Distemper Virus | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Canine distemper virus (CDV). | |
| 0.2 mL | |
| Primary | |
| Virus | |
| Purified |
| ELISA | |
| 43224 | |
| ELISA, Sandwich ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Specific for surface envelope antigen of CDV | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction