missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Distemper Virus Antibody (8-1), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-2603DL594
This item is not returnable.
View return policy
Description
Distemper Virus Monoclonal antibody specifically detects Distemper Virus in Virus samples. It is validated for ELISA, Immunoassay, ELISA, Lateral Flow
Specifications
| Distemper Virus | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Immunoassay, ELISA, Lateral Flow | |
| 2024-08-01 00:00:00 | |
| ELISA, Immunoassay, Sandwich ELISA, Lateral Flow Assay | |
| CDV, Canine Distemper Virus | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Canine distemper virus (CDV). | |
| 0.2 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction