Learn More
Lenz Laborglasintrumente Distributors for Distilling Receivers acc. to Bredt
Product Code. 10261171
Click to view available options
Standard Taper Joint:
Upper cone NS 19/26
Unit Size:
Each
Product Code. 10261171
Brand: Lenz Laborglasintrumente 05285202
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Distributors with four cones NS 14/23
  • For round bottom flasks up to 250 mL volume
  • Made of DURAN™ tubing
Specifications
Standard Taper Joint Upper cone NS 19/26