Learn More
Lenz Laborglasintrumente Distributors for Distilling Receivers acc. to Bredt
Click to view available options
Standard Taper Joint:
Upper cone NS 19/26
Unit Size:
Each
Description
- Distributors with four cones NS 14/23
- For round bottom flasks up to 250 mL volume
- Made of DURAN™ tubing
Specifications
Specifications
| Standard Taper Joint | Upper cone NS 19/26 |