Learn More
Dithieno[3,2-b:2',3'-d]thiophene, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 3593-75-7 |
| Molecular Formula | C8H4S3 |
| MDL Number | MFCD06656577 |
| Quantity | 25 g |
| SMILES | C1(C=CS2)=C2C3=C(C=CS3)S1 |
| Molecular Weight (g/mol) | 196.31 |
| Percent Purity | 98% |
| Chemical Name or Material | Dithieno[3,2-b:2',3'-d]thiophene |