Learn More
Invitrogen™ Dityrosine Monoclonal Antibody (7D4), APC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545825
Description
A 1:1,000 dilution of MA5-45825 was sufficient for detection of dityrosine in 1 μg of Dityrosine conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for dityrosine modified proteins. Does not cross-react with 3,5-dibromotyrosine or bromotyrosine modified proteins.
Dityrosine may be a natural component of protein structure, a product of environmental stress, or a product of in vitro protein modification.Dityrosine is involved in diverse reactions that produce tyrosyl radicals, which in turn may couple to yield dityrosine.
Specifications
| Dityrosine | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot, Immunocytochemistry | |
| 7D4 | |
| APC | |
| Dityrosine; Dityrosine (DT); DT; DY | |
| Synthetic Dityrosine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid | |
| Chemical |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.