Learn More
Divarinic Acid, TRC
Product Code. 30009139
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009139 5 mg 5mg
30004100 25 mg 25mg
30006564 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009139 Supplier Toronto Research Chemicals Supplier No. TRCD4944635MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4707-50-0
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.2
Synonym 6-Propyl-b-resorcylic Acid, (7CI,8CI), 2,4-Dihydroxy-6-propylbenzoic Acid, Divaric Acid
SMILES OC(C(C(O)=CC(O)=C1)=C1CCC)=O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C10H12O4/c1-2-3-6-4-7(11)5-8(12)9(6)10(13)14/h4-5,11-12H,2-3H2,1H3,(H,13,14)
Formula Weight 196.0736
Percent Purity >95%
Chemical Name or Material Divarinic Acid
Grade HPLC