missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKFZP434B168 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-465IR
This item is not returnable.
View return policy
Description
DKFZP434B168 Polyclonal antibody specifically detects DKFZP434B168 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| DKFZP434B168 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| C1orf73, chromosome 1 open reading frame 73, DKFZp434B168, Int7, integrator complex subunit 7 | |
| The immunogen this antibody was made to, maps to a region between residues 850-900 of human DKFZP434B168 using the TrEMBL entry Q9NVH2 (GeneID 25896). | |
| 0.1 mL | |
| Zinc Finger | |
| 25896 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction