missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKFZp566F084 Antibody (OTI4B9), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71779AF647
This item is not returnable.
View return policy
Description
DKFZp566F084 Monoclonal specifically detects DKFZp566F084 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| DKFZp566F084 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DPCD | |
| Full length recombinant protein of human DKFZp566F084 (NP_056263) produced in HEK293T cells. | |
| 0.1 mL | |
| 25911.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI4B9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| deleted in a mouse model of primary ciliary dyskinesia, deleted in primary ciliary dyskinesia homolog (mouse), DKFZP566F084, protein DPCD, RP11-529I10.4 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction