missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKFZp566F084 Antibody (OTI4B9), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71779PE
This item is not returnable.
View return policy
Description
DKFZp566F084 Monoclonal antibody specifically detects DKFZp566F084 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| DKFZp566F084 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 25911 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI4B9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| deleted in a mouse model of primary ciliary dyskinesia, deleted in primary ciliary dyskinesia homolog (mouse), DKFZP566F084, protein DPCD, RP11-529I10.4 | |
| Full length human recombinant protein of human DPCD(NP_056263) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction