missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dkk-1 Antibody (1D12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26620-50ul
This item is not returnable.
View return policy
Description
Dkk-1 Monoclonal antibody specifically detects Dkk-1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| Dkk-1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 1D12 | |
| Western Blot 1:500-1:5000, ELISA | |
| dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, DKK-1, hDkk-1, SKdickkopf-1 like | |
| A synthesized peptide derived from Human Dkk-1 [UniProt O94907] | |
| 50 μL | |
| Immunology, Innate Immunity, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 22943 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction