missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Dkk-1 Antibody (CL14707), Novus Biologicals™
Mouse Monoclonal Antibody
€ 317.00 - € 501.00
Specifications
| Antigen | Dkk-1 |
|---|---|
| Clone | CL14707 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043919
|
Novus Biologicals™
NBP3-26879-100ul |
100 μL |
€ 501.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043311
|
Novus Biologicals™
NBP3-26879-25ul |
25 μL |
€ 317.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dkk-1 Monoclonal antibody specifically detects Dkk-1 in Human samples. It is validated for Western BlotSpecifications
| Dkk-1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Immunology, Innate Immunity, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| PBS, pH 7.2, 40% glycerol | |
| 22943 | |
| IgG2a | |
| Protein A purified |
| CL14707 | |
| Western Blot 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, DKK-1, hDkk-1, SKdickkopf-1 like | |
| This antibody was developed against a Synthetic Peptide of O94907. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title