missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dkk-2 Antibody (994930), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6628R
This item is not returnable.
View return policy
Description
Dkk-2 Monoclonal antibody specifically detects Dkk-2 in Human samples. It is validated for Immunohistochemistry, Flow Cytometry, Immunocytochemistry
Specifications
| Dkk-2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
| 994930 | |
| Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| dickkopf (Xenopus laevis) homolog 2, dickkopf 2, dickkopf homolog 2 (Xenopus laevis), dickkopf related protein-2, dickkopf-2, dickkopf-related protein 2, DKK-2, hDkk-2 | |
| Chinese Hamster Ovary cell line, CHO-derived human Dkk-2, Met1-Ile259, Accession # NP_055236 | |
| 0.1 mL | |
| Stem Cells | |
| 27123 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction