missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DKK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590096
This item is not returnable.
View return policy
Description
Immunogen sequence: CMVCRRKKKR CHRDGMCCPS TRCNNGICIP VTESILTPHI PALDGTRHRD RNHGHYSNHD LGWQNLGRPH TKMSHIKGHE GDPCLRSSDC IEGFCCARHF WTKICKPVLH QGEVCTKQRK KGSHGLEIFQ RCDCAKGLSC KVWKDATYSS KARLHVCQKI; Positive Samples: A-549, NCI-H460.
This gene encodes a protein that is a member of the dickkopf family. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. It can act as either an agonist or antagonist of Wnt/beta-catenin signaling, depending on the cellular context and the presence of the co-factor kremen 2. Activity of this protein is also modulated by binding to the Wnt co-receptor LDL-receptor related protein 6.
Specifications
| DKK2 | |
| Polyclonal | |
| Unconjugated | |
| DKK2 | |
| dickkopf 2; dickkopf 2 homolog; Dickkopf gene 2; dickkopf homolog 1; dickkopf homolog 2; dickkopf homolog 2 (Xenopus laevis); dickkopf homolog 4; dickkopf related protein-2; dickkopf WNT signaling pathway inhibitor 2; dickkopf-2; Dickkopf-related protein 2; DKK2; DKK-2; Dkk4; hDkk-2; mDkk-2; UNQ682/PRO1316 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27123 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.84 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9UBU2 | |
| DKK2 | |
| A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human DKK2 (NP_0552361). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction