missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKK3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4E6H6, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | DKK3 |
|---|---|
| Clone | 4E6H6 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16888925
|
Proteintech
66758-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16878925
|
Proteintech
66758-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DKK3, like DKK1, DKK2, and DKK4, possesses an N-terminal signal peptide and 2 conserved cysteine-rich domains, which are separated by a linker region and contain 10 cysteine residues each. The second cysteine region has a putative lipid-binding function that may facilitate WNT/DKK interactions at the plasma membrane. The linker region contains 50 to 55 amino acids in DKK1, DKK2, and DKK4, whereas in DKK3 it contains only 12 amino acids. All DKKs have several potential sites for cleavage by furin-type proteases. Northern blot analysis revealed wide expression of the DKK3 transcript, with highest expression in heart, brain, and spinal cord. In situ hybridization reveals highest expression in mouse brain, eye, and heart.Specifications
| DKK3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Dickkopf 3, Dickkopf related protein 3, Dkk 3, DKK3, hDkk 3, REIC, RIG | |
| DKK3 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 4E6H6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Pig, Mouse, Rat | |
| Q9QUN9, Q9UBP4 | |
| 171548, 27122, 50781, 664653 | |
| DKK3 Fusion Protein Ag23615 | |
| Primary | |
| -20°C | |
| DKK3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title